Gene Transcripts

Gene Symbol
Olr1334
Entrez Gene ID
300634
Gene Name
olfactory receptor 1334
Gene Aliases
Chromosome Location
Chr. 8: 43848058 - 43848993 on Build Rnor_6.0
UniGene ID
Rn.143467
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000478.1 NP_001000478.1 1 484

View Full Product Details