Gene Transcripts

Gene Symbol
Cyp2b10
Entrez Gene ID
13088
Gene Name
cytochrome P450, family 2, subfamily b, polypeptide 10
Gene Aliases
Cyp2b, Cyp2b20, p16
Chromosome Location
Chr. 7: 25897658 - 25926624 on Build GRCm38
UniGene ID
Mm.218749
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009999.4 NP_034129.1 2 350
GenBank mRNA AF128849.1 - 2 332
AF129405.1 - 1 146
AI304065.1 - 2 304
AK028103.1 - 2 350
BC060973.1 - 2 344 30301385
M21856.1 - 2 332
X99715.1 - 2 328

View Full Product Details