Gene Transcripts

Gene Symbol
Rasip1
Entrez Gene ID
69903
Gene Name
Ras interacting protein 1
Gene Aliases
1110025D03Rik, 2610025P08Rik, AI853551, Rain
Chromosome Location
Chr. 7: 45627537 - 45639092 on Build GRCm38
UniGene ID
Mm.181396
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028544.1 NP_082820.1 4 993
GenBank mRNA AK033276.1 - 2 281
AK156602.1 - 4 1005
BB646451.1 - 2 602
BC150766.1 - 4 993 9087677

View Full Product Details