Gene Transcripts

Gene Symbol
Nipal3
Entrez Gene ID
74552
Gene Name
NIPA-like domain containing 3
Gene Aliases
9130020G22Rik, AI480660, Npal3
Chromosome Location
Chr. 4: 135447136 - 135494806 on Build GRCm38
UniGene ID
Mm.26548
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028995.3 NP_083271.1 5 706
GenBank mRNA AK018640.1 - 5 683
AK049696.1 - 5 741
AK053937.1 - 709
AK082376.1 - 708
AK087811.1 - 5 709
AK138843.1 - 5 707
BC057168.1 - 5 646 4973200

View Full Product Details