Gene Transcripts
- Gene Symbol
- Olfr968
- Entrez Gene ID
- 258605
- Gene Name
- olfactory receptor 968
- Gene Aliases
- MOR171-15
- Chromosome Location
- Chr. 9: 39771772 - 39772798 on Build GRCm38
- UniGene ID
- Mm.271458
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_146612.2 |
NP_666823.1
|
1 |
768 |
|
View Full Product Details