Gene Transcripts

Gene Symbol
Olfr235
Entrez Gene ID
258681
Gene Name
olfactory receptor 235
Gene Aliases
MOR214-3, Olfr232, Olfr232-ps1, Olfr235-ps1, Olfr245, Olfr245-ps1
Chromosome Location
Chr. 19: 12268232 - 12269170 on Build GRCm38
UniGene ID
Mm.271522
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146686.2 NP_666897.2 1 109
GenBank mRNA BC132238.1 - 1 179
BC132388.1 - 1 179 40130665

View Full Product Details