Gene Transcripts

Gene Symbol
NDUFV2
Entrez Gene ID
4729
Gene Name
NADH:ubiquinone oxidoreductase core subunit V2
Gene Aliases
CI-24k
Chromosome Location
Chr. 18: 9102623 - 9134345 on Build GRCh38
UniGene ID
Hs.464572
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK291865.1 - 2 86
AK312129.1 - 2 108
BC071689.1 - 2 80 4372795
BG742948.1 - 2 190
CR456928.1 - 2 76
M22538.1 - 2 94

View Full Product Details