Gene Transcripts

Gene Symbol
FUT2
Entrez Gene ID
2524
Gene Name
fucosyltransferase 2
Gene Aliases
B12QTL1, SE, SEC2, Se2, sej
Chromosome Location
Chr. 19: 48695971 - 48705934 on Build GRCh38
UniGene ID
Hs.579928
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000511.5 NP_000502.4 2 265
NM_001097638.2 NP_001091107.1 2 255
GenBank mRNA AK290277.1 - 2 251
AY751739.1 - 1 144
BC001899.1 - 2 205
BC121066.1 - 2 187
BE266792.1 - 2 187
BT006657.1 - 1 144
D87942.1 - 2 255
DA918525.1 - 2 255

View Full Product Details