Gene Transcripts

Gene Symbol
SP5
Entrez Gene ID
389058
Gene Name
Sp5 transcription factor
Gene Aliases
Chromosome Location
Chr. 2: 170714725 - 170717988 on Build GRCh38
UniGene ID
Hs.368802
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003845.2 NP_001003845.1 2 1886
XM_005246542.4 XP_005246599.1 2 3013
XM_011511159.2 XP_011509461.1 2 3049
GenBank mRNA AB096175.1 - 2 1881

View Full Product Details