Gene Transcripts

Gene Symbol
ZNF592
Entrez Gene ID
9640
Gene Name
zinc finger protein 592
Gene Aliases
CAMOS, SCAR5
Chromosome Location
Chr. 15: 84748556 - 84806432 on Build GRCh38
UniGene ID
Hs.79347
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014630.2 NP_055445.2 7 2945
XM_005254996.3 XP_005255053.1 6 2846
XM_011522246.2 XP_011520548.1 7 3015
XM_011522247.2 XP_011520549.1 6 3018
XM_011522248.2 XP_011520550.1 7 2976
XM_017022734.1 XP_016878223.1 5 2726
GenBank mRNA BC094688.1 - 7 2897
BC112232.1 - 5 2679 8327700
BC112234.1 - 5 2680 8327702
BC143600.1 - 5 2670
BG757479.1 - 1 33
BQ217254.1 - 4 658
D86966.1 - 7 3179

View Full Product Details