Gene Transcripts

Gene Symbol
C5AR2
Entrez Gene ID
27202
Gene Name
complement component 5a receptor 2
Gene Aliases
C5L2, GPF77, GPR77
Chromosome Location
Chr. 19: 47331614 - 47342080 on Build GRCh38
UniGene ID
Hs.534412
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271749.1 NP_001258678.1 2 1194
NM_001271750.1 NP_001258679.1 2 1117
NM_018485.2 NP_060955.1 2 1156
XM_011526736.2 XP_011525038.1 3 1831
GenBank mRNA AB038237.1 - 2 1047
AK313994.1 - 2 1194
BC067477.1 - 1 995 6971811
BC067478.1 - 1 995 6971812
BC067479.1 - 1 995 6971813
BC067480.1 - 1 995 6971814

View Full Product Details