Gene Transcripts

Gene Symbol
SH3BP1
Entrez Gene ID
23616
Gene Name
SH3 domain binding protein 1
Gene Aliases
ARHGAP43
Chromosome Location
Chr. 22: 37639677 - 37656043 on Build GRCh38
UniGene ID
Hs.601143
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018957.3 NP_061830.3 7 718
GenBank mRNA AK124370.1 - 8 1545
AK126556.1 - 6 1413
AK301163.1 - 7 700
AY364241.1 - 4 415
BC008282.1 - 5 511 3639782
CR456576.1 - 7 637

View Full Product Details