Gene Transcripts

Gene Symbol
KCNT2
Entrez Gene ID
343450
Gene Name
potassium sodium-activated channel subfamily T member 2
Gene Aliases
KCa4.2, SLICK, SLO2.1
Chromosome Location
Chr. 1: 196225779 - 196608905 on Build GRCh38
UniGene ID
Hs.657046
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001287819.1 NP_001274748.1 1, 2 225
NM_001287820.1 NP_001274749.1 1, 2 225
NM_198503.3 NP_940905.2 1, 2 225
XM_006711294.2 XP_006711357.1 1, 2 699
XM_006711295.2 XP_006711358.1 1, 2 699
XM_011509482.2 XP_011507784.1 1, 2 699
XM_011509483.2 XP_011507785.1 1, 2 699
XM_017001178.1 XP_016856667.1 1, 2 699
XM_017001179.1 XP_016856668.1 1, 2 699
XM_017001180.1 XP_016856669.1 1, 2 699
XM_017001182.1 XP_016856671.1 1, 2 699
XM_017001184.1 XP_016856673.1 1, 2 699
XM_017001185.1 XP_016856674.1 1, 2 699
GenBank mRNA AK131498.1 - 1, 2 354
AK309729.1 - 1, 2 214
AY359444.1 - 1, 2 163
BC103947.1 - 1, 2 153
BC103948.1 - 1, 2 153 40009146
BC103949.1 - 1, 2 153 40009147
BC103950.1 - 1, 2 153 40009148
BX647852.1 - 1, 2 206
DA799907.1 - 1, 2 225
EU251076.1 - 1, 2 103

View Full Product Details