Gene Transcripts
- Gene Symbol
- NDUFV2
- Entrez Gene ID
- 4729
- Gene Name
- NADH:ubiquinone oxidoreductase core subunit V2
- Gene Aliases
- CI-24k
- Chromosome Location
- Chr. 18: 9102623 - 9134345 on Build GRCh38
- UniGene ID
- Hs.464572
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_021074.4 |
NP_066552.2
|
2, 3 |
228 |
|
XM_017025782.1 |
XP_016881271.1
|
2, 3 |
418 |
|
GenBank mRNA |
AK291865.1 |
- |
2, 3 |
124 |
|
AK312129.1 |
- |
2, 3 |
146 |
|
BC001632.1 |
- |
2, 3 |
138 |
3536345 |
BC017487.1 |
- |
2, 3 |
136 |
3537815 |
BC071689.1 |
- |
2, 3 |
118 |
4372795 |
BG742948.1 |
- |
2, 3 |
228 |
|
CB121843.1 |
- |
2, 3 |
181 |
|
CR456928.1 |
- |
2, 3 |
114 |
|
M22538.1 |
- |
2, 3 |
132 |
|
View Full Product Details