Gene Transcripts

Gene Symbol
FGFRL1
Entrez Gene ID
53834
Gene Name
fibroblast growth factor receptor-like 1
Gene Aliases
FGFR5, FHFR
Chromosome Location
Chr. 4: 1011822 - 1026898 on Build GRCh38
UniGene ID
Hs.193326
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004356.2 NP_001004356.1 7 2000
NM_001004358.1 NP_001004358.1 7 1883
NM_021923.3 NP_068742.2 6 1873
XM_011513486.1 XP_011511788.1 7 2004
GenBank mRNA AF312678.1 - 1977
AJ277437.1 - 6 1873
AK022597.1 - 1 785
AY358303.1 - 1998
BC015228.1 - 3 1284
BC036769.1 - 7 2012 5744684

View Full Product Details