Gene Transcripts

Gene Symbol
ALDH8A1
Entrez Gene ID
64577
Gene Name
aldehyde dehydrogenase 8 family member A1
Gene Aliases
ALDH12, DJ352A20.2
Chromosome Location
Chr. 6: 134917390 - 134950122 on Build GRCh38
UniGene ID
Hs.486520
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001193480.1 NP_001180409.1 2, 3 363
NM_022568.3 NP_072090.1 2, 3 363
NM_170771.2 NP_739577.1 2, 3 363
GenBank mRNA AF303134.1 - 2, 3 354
AK074266.1 - 2, 3 356
AK222848.1 - 2, 3 356
AK290784.1 - 2, 3 310
AK298325.1 - 2, 3 325
BC113862.1 - 2, 3 333 40033493
BC114473.1 - 2, 3 330 40033488
DA635162.1 - 2, 3 363

View Full Product Details