Gene Transcripts

Gene Symbol
MLLT6
Entrez Gene ID
4302
Gene Name
MLLT6, PHD finger domain containing
Gene Aliases
AF17
Chromosome Location
Chr. 17: 38705620 - 38729803 on Build GRCh38
UniGene ID
Hs.91531
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005937.3 NP_005928.2 10, 11 1672
XM_006721911.2 XP_006721974.1 10, 11 1594
XM_017024648.1 XP_016880137.1 5, 6 1197
XM_017024649.1 XP_016880138.1 10, 11 1672
XM_017024650.1 XP_016880139.1 10, 11 1672
GenBank mRNA AB209633.1 - 10, 11 1647
BC007859.2 - 1 23
U07932.1 - 10, 11 1659

View Full Product Details