Gene Transcripts

Gene Symbol
PRKCG
Entrez Gene ID
5582
Gene Name
protein kinase C gamma
Gene Aliases
PKC-gamma, PKCC, PKCG, SCA14
Chromosome Location
Chr. 19: 53882184 - 53907647 on Build GRCh38
UniGene ID
Hs.631564
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001316329.1 NP_001303258.1 6 856
NM_002739.4 NP_002730.1 6 856
GenBank mRNA AB209671.1 - 6 575
AF345987.1 - 6 740
AK296412.1 - 6 864
AK296434.1 - 6 843
AK302958.1 - 6 841
AK303741.1 - 4 263
AK312302.1 - 6 818
BC047876.1 - 6 731 5764695
M13977.1 - 6 740
Z15114.1 - 2 64

View Full Product Details