Gene Transcripts

Gene Symbol
SLC27A3
Entrez Gene ID
11000
Gene Name
solute carrier family 27 member 3
Gene Aliases
ACSVL3, FATP3, VLCS-3
Chromosome Location
Chr. 1: 153774989 - 153780157 on Build GRCh38
UniGene ID
Hs.438723
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317929.1 NP_001304858.1 1 151
NM_024330.1 NP_077306.1 1 151
GenBank mRNA AJ577572.1 - 1 444
AK027499.1 - 1 55
AK074134.1 - 1 124
AK075377.1 - 1 144
AK123997.1 - 1 133
AK125102.1 - 1 1089
AY358409.1 - 1 139
BC003041.1 - 1 151 2822875
BC009916.2 - 1 151 2822875
BC029792.1 - 1 19 5195359

View Full Product Details