Gene Transcripts
- Gene Symbol
- TNIP2
- Entrez Gene ID
- 79155
- Gene Name
- TNFAIP3 interacting protein 2
- Gene Aliases
- ABIN2, FLIP1, KLIP
- Chromosome Location
- Chr. 4: 2741660 - 2756376 on Build GRCh38
- UniGene ID
- Hs.368551
- Species
- Homo sapiens
- Gene Symbol
- LOC100132831
- Entrez Gene ID
- 100132831
- Gene Name
- A20-binding inhibitor of NF-kappaB activation 2 pseudogene
- Gene Aliases
- Chromosome Location
- Chr. X: 40831217 - 40833196 on Build GRCh38
- UniGene ID
- Hs.729923
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027761.1 |
|
1 |
711 |
|
View Full Product Details