Gene Transcripts
- Gene Symbol
- CHPF
- Entrez Gene ID
- 79586
- Gene Name
- chondroitin polymerizing factor
- Gene Aliases
- CHSY2, CSS2
- Chromosome Location
- Chr. 2: 219538947 - 219543765 on Build GRCh38
- UniGene ID
- Hs.516711
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001195731.1 |
NP_001182660.1
|
3, 4 |
938 |
|
NM_024536.5 |
NP_078812.2
|
3, 4 |
1294 |
|
XM_011511838.2 |
XP_011510140.1
|
2, 3 |
825 |
|
GenBank mRNA |
AB086063.1 |
- |
3, 4 |
1128 |
|
AB095813.1 |
- |
3, 4 |
1303 |
|
AK302124.1 |
- |
3, 4 |
938 |
|
AL136814.1 |
- |
2, 3 |
441 |
|
AY358403.1 |
- |
3, 4 |
1294 |
|
BC021223.2 |
- |
3, 4 |
1294 |
4128192 |
BC023531.2 |
- |
3, 4 |
1245 |
4123041 |
View Full Product Details