Gene Transcripts

Gene Symbol
MRPL24
Entrez Gene ID
79590
Gene Name
mitochondrial ribosomal protein L24
Gene Aliases
L24mt, MRP-L18, MRP-L24
Chromosome Location
Chr. 1: 156737302 - 156741267 on Build GRCh38
UniGene ID
Hs.418233
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024540.3 NP_078816.2 3, 4 415
NM_145729.2 NP_663781.1 3, 4 456
XM_011509981.2 XP_011508283.1 3, 4 526
XM_011509982.2 XP_011508284.1 3, 4 424
GenBank mRNA AK024570.1 - 3, 4 413
AK222500.1 - 3, 4 413
AK223066.1 - 3, 4 413
BC012440.1 - 3, 4 397 3863359
BC016700.1 - 3, 4 415 4108596
BG706700.1 - 3, 4 420
CR457329.1 - 2, 3 276

View Full Product Details