Gene Transcripts

Gene Symbol
VPS37B
Entrez Gene ID
79720
Gene Name
VPS37B, ESCRT-I subunit
Gene Aliases
Chromosome Location
Chr. 12: 122865328 - 122896165 on Build GRCh38
UniGene ID
Hs.507162
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024667.2 NP_078943.1 2 283
XM_005253622.3 XP_005253679.1 3 2998
XM_006719602.3 XP_006719665.1 3 319
XM_011538743.2 XP_011537045.1 3 424
XM_011538745.2 XP_011537047.1 2 2814
XM_017019970.1 XP_016875459.1 3 428
GenBank mRNA AK022812.1 - 2 283
BC005882.2 - 2 221 3533384

View Full Product Details