Gene Transcripts

Gene Symbol
ZDHHC11B
Entrez Gene ID
653082
Gene Name
zinc finger DHHC-type containing 11B
Gene Aliases
DHHC-11B
Chromosome Location
Chr. 5: 710355 - 784749 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017010110.1 XP_016865599.1 8, 9 1277
XM_017010111.1 XP_016865600.1 7, 8 1104
XM_017010112.1 XP_016865601.1 7, 8 1681
XM_017010113.1 XP_016865602.1 8, 9 1803
XM_017010114.1 XP_016865603.1 8, 9 4564
XM_017010115.1 XP_016865604.1 7, 8 1629
XM_017010116.1 XP_016865605.1 6, 7 2029
XM_017010117.1 XP_016865606.1 6, 7 1864
XM_017010123.1 XP_016865612.1 6, 7 1778
XM_017010124.1 XP_016865613.1 7, 8 1788
XM_017010125.1 XP_016865614.1 3, 4 1353

Gene Symbol
ZDHHC11
Entrez Gene ID
79844
Gene Name
zinc finger DHHC-type containing 11
Gene Aliases
ZNF399
Chromosome Location
Chr. 5: 795605 - 863634 on Build GRCh38
UniGene ID
Hs.654874
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024786.2 NP_079062.1 4, 5 1012
XM_017009869.1 XP_016865358.1 5, 6 893
XM_017009870.1 XP_016865359.1 7, 8 2704
XM_017009871.1 XP_016865360.1 7, 8 2653
XM_017009872.1 XP_016865361.1 5, 6 728
GenBank mRNA AK023215.1 - 4, 5 1012
BC032000.1 - 4, 5 631 4750082

View Full Product Details