Gene Transcripts

Gene Symbol
AEBP1
Entrez Gene ID
165
Gene Name
AE binding protein 1
Gene Aliases
ACLP
Chromosome Location
Chr. 7: 44104320 - 44114565 on Build GRCh38
UniGene ID
Hs.439463
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001129.4 NP_001120.3 2 602
XM_011515162.1 XP_011513464.1 2 643
GenBank mRNA AB209744.1 - 2 556
AF053944.1 - 2 436
BC038588.1 - 2 602 5764769

View Full Product Details