Gene Transcripts

Gene Symbol
QSER1
Entrez Gene ID
79832
Gene Name
glutamine and serine rich 1
Gene Aliases
Chromosome Location
Chr. 11: 32892826 - 32980270 on Build GRCh38
UniGene ID
Hs.369368
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001076786.2 NP_001070254.1 5, 6 4459
XM_017018327.1 XP_016873816.1 5, 6 4811
XM_017018328.1 XP_016873817.1 5, 6 4811
XM_017018329.1 XP_016873818.1 3, 4 5264
XM_017018330.1 XP_016873819.1 4, 5 4209
GenBank mRNA AK025577.1 - 2, 3 1186
AK123858.1 - 2, 3 2521
AK125902.1 - 2, 3 2007
AK126023.1 - 6, 7 3763
AL834141.1 - 2, 3 3155
BC112935.1 - 5, 6 4459

View Full Product Details