Gene Transcripts

Gene Symbol
NOL10
Entrez Gene ID
79954
Gene Name
nucleolar protein 10
Gene Aliases
PQBP5
Chromosome Location
Chr. 2: 10570766 - 10689991 on Build GRCh38
UniGene ID
Hs.222494
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001261392.1 NP_001248321.1 3 275
NM_001261394.1 NP_001248323.1 3 275
NM_024894.3 NP_079170.2 3 275
NR_048552.1 2 229
XM_011510398.2 XP_011508700.1 3 279
XM_011510399.2 XP_011508701.1 2 217
XM_011510400.2 XP_011508702.1 3 263
XM_011510402.2 XP_011508704.1 3 263
XM_017004990.1 XP_016860479.1 1 17
GenBank mRNA AK024000.1 - 3 253
AK024137.1 - 3 260
AK290680.1 - 3 239
AK297169.1 - 3 263
AK299617.1 - 2 217
BC005125.2 - 3 255 3841643
DC346574.1 - 3 275

View Full Product Details