Gene Transcripts
- Gene Symbol
- JADE1
- Entrez Gene ID
- 79960
- Gene Name
- jade family PHD finger 1
- Gene Aliases
- PHF17
- Chromosome Location
- Chr. 4: 128809623 - 128875224 on Build GRCh38
- UniGene ID
- Hs.12420
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001287437.1 |
NP_001274366.1
|
3, 4 |
427 |
|
NM_001287439.1 |
NP_001274368.1
|
3, 4 |
545 |
|
NM_001287440.1 |
NP_001274369.1
|
3, 4 |
342 |
|
NM_001287441.1 |
NP_001274370.1
|
3, 4 |
306 |
|
NM_001287442.1 |
NP_001274371.1
|
3, 4 |
299 |
|
NM_001287443.1 |
NP_001274372.1
|
3, 4 |
303 |
|
NM_024900.4 |
NP_079176.2
|
3, 4 |
427 |
|
NM_199320.3 |
NP_955352.1
|
3, 4 |
427 |
|
XM_005263232.1 |
XP_005263289.1
|
3, 4 |
269 |
|
XM_017008626.1 |
XP_016864115.1
|
3, 4 |
384 |
|
XM_017008627.1 |
XP_016864116.1
|
3, 4 |
431 |
|
GenBank mRNA |
AF520952.1 |
- |
3, 4 |
324 |
|
AK027620.1 |
- |
3, 4 |
347 |
|
AK127326.1 |
- |
3, 4 |
235 |
|
AK304235.1 |
- |
3, 4 |
260 |
|
AK304815.1 |
- |
3, 4 |
241 |
|
BC032376.1 |
- |
3, 4 |
261 |
5111727 |
DA478923.1 |
- |
3, 4 |
341 |
|
DA714593.1 |
- |
3, 4 |
299 |
|
DB085413.1 |
- |
3, 4 |
303 |
|
DB293142.1 |
- |
3, 4 |
281 |
|
DB451567.1 |
- |
3, 4 |
426 |
|
DC308588.1 |
- |
3, 4 |
536 |
|
View Full Product Details