Gene Transcripts
- Gene Symbol
- ZNF767P
- Entrez Gene ID
- 79970
- Gene Name
- zinc finger family member 767, pseudogene
- Gene Aliases
- ZNF767
- Chromosome Location
- Chr. 7: 149547154 - 149624790 on Build GRCh38
- UniGene ID
- Hs.520785
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027788.1 |
|
4 |
791 |
|
GenBank mRNA |
AK022762.1 |
- |
4 |
728 |
|
AK295771.1 |
- |
5 |
756 |
|
View Full Product Details