Gene Transcripts

Gene Symbol
CTC1
Entrez Gene ID
80169
Gene Name
CTS telomere maintenance complex component 1
Gene Aliases
AAF-132, AAF132, C17orf68, CRMCC, tmp494178
Chromosome Location
Chr. 17: 8224821 - 8248095 on Build GRCh38
UniGene ID
Hs.156055
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025099.5 NP_079375.3 13 2428
NR_046431.1 13 2323
XM_006721577.3 XP_006721640.1 12 2283
XM_006721578.3 XP_006721641.1 13 2412
XM_011524010.2 XP_011522312.1 13 2307
XM_011524011.2 XP_011522313.1 12 2177
GenBank mRNA AK025823.1 - 3 571
AK091077.1 - 13 2379
AK225375.1 - 3 571
AK310636.1 - 2 597
AL831955.1 - 13 2270
BC026057.2 - 4 569
BC110373.1 - 5 795
BC111783.1 - 13 2379 40054070
BX406692.2 - 1 181
DQ451688.1 - 13 2369
FJ790670.1 - 13 2369

View Full Product Details