Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001102371.1 | NP_001095841.1 | 9 | 3241 | |
NM_024955.5 | NP_079231.4 | 9 | 3280 | ||
XM_017028941.1 | XP_016884430.1 | 8 | 2913 | ||
XM_017028942.1 | XP_016884431.1 | 8 | 3010 | ||
GenBank mRNA | AK091053.1 | - | 4 | 2172 | |
BC027716.2 | - | 9 | 3259 | 4127473 |