Gene Transcripts

Gene Symbol
PIF1
Entrez Gene ID
80119
Gene Name
PIF1 5'-to-3' DNA helicase
Gene Aliases
C15orf20, PIF
Chromosome Location
Chr. 15: 64815630 - 64827173 on Build GRCh38
UniGene ID
Hs.112160
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286496.1 NP_001273425.1 4 824
NM_001286497.1 NP_001273426.1 4 800
NM_001286499.1 NP_001273428.1 4 824
NM_025049.3 NP_079325.2 4 800
XM_011522083.2 XP_011520385.1 4 916
XM_011522084.2 XP_011520386.1 4 748
GenBank mRNA AB185926.1 - 4 800
AB185927.1 - 4 800
AK026345.1 - 3 843
AY498716.1 - 3 724
BC137503.1 - 4 738 9021503
BC137504.1 - 4 738 9021504
DQ437529.1 - 3 705
EU084033.1 - 4 795

View Full Product Details