Gene Transcripts

Gene Symbol
SPACA1
Entrez Gene ID
81833
Gene Name
sperm acrosome associated 1
Gene Aliases
SAMP32
Chromosome Location
Chr. 6: 88046907 - 88066832 on Build GRCh38
UniGene ID
Hs.161241
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030960.2 NP_112222.1 1 269
GenBank mRNA AF203447.1 - 1 224
BC029488.1 - 1 269 5270534

View Full Product Details