Gene Transcripts

Gene Symbol
CDT1
Entrez Gene ID
81620
Gene Name
chromatin licensing and DNA replication factor 1
Gene Aliases
DUP, RIS2
Chromosome Location
Chr. 16: 88803778 - 88809258 on Build GRCh38
UniGene ID
Hs.122908
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030928.3 NP_112190.2 10 2112
GenBank mRNA AB053172.1 - 10 2112
BC000137.2 - 10 2074 3048353
BC008860.2 - 10 2005

View Full Product Details