Gene Transcripts

Gene Symbol
TMEM245
Entrez Gene ID
23731
Gene Name
transmembrane protein 245
Gene Aliases
C9orf5, CG-2, CG2
Chromosome Location
Chr. 9: 109015135 - 109119963 on Build GRCh38
UniGene ID
Hs.308074
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032012.3 NP_114401.2 4 911
XM_011518446.2 XP_011516748.1 4 929
XM_011518447.1 XP_011516749.1 4 911
XM_011518451.1 XP_011516753.1 4 911
XM_011518452.1 XP_011516754.1 4 911
XM_017014571.1 XP_016870060.1 4 911
XM_017014572.1 XP_016870061.1 4 911
GenBank mRNA AF153415.1 - 4 911
AK299952.1 - 4 893
AL833960.1 - 3 218
BP360669.1 - 1 58

View Full Product Details