Gene Transcripts

Gene Symbol
SLC37A3
Entrez Gene ID
84255
Gene Name
solute carrier family 37 member 3
Gene Aliases
Chromosome Location
Chr. 7: 140333752 - 140404352 on Build GRCh38
UniGene ID
Hs.446021
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001287498.1 NP_001274427.1 14 1836
NM_032295.3 NP_115671.1 12 1582
NM_207113.2 NP_996996.1 15 1884
XM_011516626.2 XP_011514928.1 16 2229
XM_011516627.2 XP_011514929.1 15 2180
XM_017012712.1 XP_016868201.1 17 2357
XM_017012713.1 XP_016868202.1 14 2074
XM_017012714.1 XP_016868203.1 15 2154
XM_017012715.1 XP_016868204.1 15 2136
XM_017012716.1 XP_016868205.1 12 1838
XM_017012717.1 XP_016868206.1 13 1932
GenBank mRNA AK074823.1 - 15 1845
AK126589.1 - 3 1028
AL136583.1 - 12 1489
BC028380.1 - 15 1834 4811814
BC046567.1 - 14 1788 5246993

View Full Product Details