Gene Transcripts

Gene Symbol
USP30
Entrez Gene ID
84749
Gene Name
ubiquitin specific peptidase 30
Gene Aliases
Chromosome Location
Chr. 12: 109023089 - 109088026 on Build GRCh38
UniGene ID
Hs.486434
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301175.1 NP_001288104.1 5 633
NM_032663.4 NP_116052.2 2 259
XM_005253962.3 XP_005254019.1 2 245
XM_005253965.4 XP_005254022.1 2 253
XM_006719653.3 XP_006719716.1 3 332
XM_017020048.1 XP_016875537.1 2 254
XM_017020049.1 XP_016875538.1 2 245
XM_017020050.1 XP_016875539.1 2 354
XM_017020051.1 XP_016875540.1 3 210
XM_017020052.1 XP_016875541.1 2 256
XM_017020053.1 XP_016875542.1 2 169
GenBank mRNA AJ586136.1 - 2 150
AK027820.1 - 2 224
AK097830.1 - 5 633
BC004868.2 - 2 249 3833242

View Full Product Details