Gene Transcripts

Gene Symbol
KLHL22
Entrez Gene ID
84861
Gene Name
kelch like family member 22
Gene Aliases
KELCHL
Chromosome Location
Chr. 22: 20441519 - 20496249 on Build GRCh38
UniGene ID
Hs.517419
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032775.3 NP_116164.2 3 417
XM_017029019.1 XP_016884508.1 3 713
XM_017029020.1 XP_016884509.1 4 1892
XM_017029021.1 XP_016884510.1 3 346
XM_017029025.1 XP_016884514.1 2 65
GenBank mRNA AK027266.1 - 3 356
AK222850.1 - 3 323
AK290669.1 - 3 329
BC015923.1 - 3 369 4562420
CR456352.1 - 2 294
DA381792.1 - 3 417

View Full Product Details