Gene Transcripts

Gene Symbol
FAM222A
Entrez Gene ID
84915
Gene Name
family with sequence similarity 222 member A
Gene Aliases
C12orf34
Chromosome Location
Chr. 12: 109714382 - 109770509 on Build GRCh38
UniGene ID
Hs.661785
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032829.2 NP_116218.2 3 2213
XM_006719654.2 XP_006719717.1 3 1799
XM_017020055.1 XP_016875544.1 3 2183
GenBank mRNA AK027627.1 - 3 1817
AK074806.1 - 1 1472
BC007844.2 - 2 278
BC020193.1 - 3 2210
BC037221.1 - 2 1688 5197135

View Full Product Details