Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001102603.1 | NP_001096073.1 | 5 | 513 | |
NM_001322125.1 | NP_001309054.1 | 4 | 396 | ||
NM_001322126.1 | NP_001309055.1 | 5 | 537 | ||
NM_001322128.1 | NP_001309057.1 | 5 | 521 | ||
NM_001322129.1 | NP_001309058.1 | 4 | 616 | ||
NM_001322130.1 | NP_001309059.1 | 5 | 745 | ||
NM_001322131.1 | NP_001309060.1 | 4 | 620 | ||
NM_001322132.1 | NP_001309061.1 | 5 | 509 | ||
NM_001322133.1 | NP_001309062.1 | 5 | 533 | ||
NM_001322134.1 | NP_001309063.1 | 5 | 537 | ||
NM_001322135.1 | NP_001309064.1 | 4 | 392 | ||
NM_001322136.1 | NP_001309065.1 | 3 | 249 | ||
NM_001322137.1 | NP_001309066.1 | 6 | 569 | ||
NM_001322138.1 | NP_001309067.1 | 5 | 452 | ||
NM_001322139.1 | NP_001309068.1 | 7 | 666 | ||
NM_033288.3 | NP_150630.2 | 4 | 396 | ||
NM_198893.2 | NP_942596.1 | 5 | 481 | ||
XM_017027445.1 | XP_016882934.1 | 2 | 745 | ||
XM_017027446.1 | XP_016882935.1 | 4 | 706 | ||
XM_017027447.1 | XP_016882936.1 | 2 | 745 | ||
XM_017027448.1 | XP_016882937.1 | 4 | 854 | ||
GenBank mRNA | AB046831.1 | - | 1 | 866 | |
AF277623.1 | - | 4 | 404 | ||
BC000807.1 | - | 5 | 506 | ||
BC094880.1 | - | 5 | 398 | 3066532 | |
BU430880.1 | - | 5 | 405 | ||
DA230397.1 | - | 5 | 575 | ||
DB078640.1 | - | 4 | 396 |