Gene Transcripts

Gene Symbol
CELF6
Entrez Gene ID
60677
Gene Name
CUGBP, Elav-like family member 6
Gene Aliases
BRUNOL6
Chromosome Location
Chr. 15: 72284727 - 72320184 on Build GRCh38
UniGene ID
Hs.348342
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001172684.1 NP_001166155.1 12 2480
NM_001172685.1 NP_001166156.1 11 2253
NM_052840.4 NP_443072.3 13 2799
GenBank mRNA AK124858.1 - 12 2353
AK131098.1 - 16 3838
BC030835.1 - 13 2501 4793742

View Full Product Details