Gene Transcripts

Gene Symbol
PNMA5
Entrez Gene ID
114824
Gene Name
paraneoplastic Ma antigen family member 5
Gene Aliases
Chromosome Location
Chr. X: 152988824 - 152994458 on Build GRCh38
UniGene ID
Hs.573567
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001103150.1 NP_001096620.1 2 596
NM_001103151.1 NP_001096621.1 3 496
NM_001184924.1 NP_001171853.1 4 581
NM_052926.2 NP_443158.1 2 582
XM_017029252.1 XP_016884741.1 4 898
XM_017029253.1 XP_016884742.1 3 560
GenBank mRNA AB067521.1 - 2 579
AK295446.1 - 3 460
AK316104.1 - 3 465
AY121806.1 - 2 579
BC101111.1 - 2 440 40011824
BC101112.2 - 2 426 40011826
BC101113.1 - 2 426 40011827
DC329825.1 - 3 460

View Full Product Details