Gene Transcripts

Gene Symbol
ATP6V1E2
Entrez Gene ID
90423
Gene Name
ATPase H+ transporting V1 subunit E2
Gene Aliases
ATP6E1, ATP6EL2, ATP6V1EL2, VMA4
Chromosome Location
Chr. 2: 46511835 - 46542557 on Build GRCh38
UniGene ID
Hs.659656
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318063.1 NP_001304992.1 5 1295
XM_005264633.2 XP_005264690.1 5 2026
XM_005264634.2 XP_005264691.1 4 1919
XM_011533148.2 XP_011531450.1 6 2725
XM_011533149.2 XP_011531451.1 5 2618
XM_011533150.2 XP_011531452.1 7 2926
XM_011533151.2 XP_011531453.1 6 2080
XM_011533152.2 XP_011531454.1 6 2797
XM_011533153.2 XP_011531455.1 5 1973
XM_017005225.1 XP_016860714.1 7 1846
XM_017005226.1 XP_016860715.1 5 2098
GenBank mRNA AB074759.1 - 2 956
AK058055.1 - 2 729
BC008981.1 - 2 956 3456620
BC034808.1 - 2 1695 5167103

View Full Product Details