Gene Transcripts

Gene Symbol
CPNE4
Entrez Gene ID
131034
Gene Name
copine 4
Gene Aliases
COPN4, CPN4
Chromosome Location
Chr. 3: 131533560 - 132285702 on Build GRCh38
UniGene ID
Hs.199877
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289112.1 NP_001276041.1 2 357
NM_130808.2 NP_570720.1 2 985
NM_153429.1 NP_702907.1 3 345
XM_017005693.1 XP_016861182.1 5 1738
XM_017005694.1 XP_016861183.1 1 109
XM_017005695.1 XP_016861184.1 2 378
GenBank mRNA AF465770.1 - 5 685
AF465771.1 - 3 345
AK056175.1 - 2 457
AK128117.1 - 6 2235
AK295557.1 - 2 272
BC014396.1 - 2 543 3162811
BC075046.2 - 2 134 30915576
DA156722.1 - 2 443
DC330179.1 - 2 357

View Full Product Details