Gene Transcripts
- Gene Symbol
- SEMA6B
- Entrez Gene ID
- 10501
- Gene Name
- semaphorin 6B
- Gene Aliases
- SEM-SEMA-Y, SEMA-VIB, SEMAN, semaZ
- Chromosome Location
- Chr. 19: 4542588 - 4581484 on Build GRCh38
- UniGene ID
- Hs.465642
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_032108.3 |
NP_115484.2
|
2, 3 |
386 |
|
XM_011527639.2 |
XP_011525941.1
|
2, 3 |
435 |
|
XM_011527640.2 |
XP_011525942.1
|
2, 3 |
178 |
|
XM_011527641.1 |
XP_011525943.1
|
2, 3 |
435 |
|
GenBank mRNA |
AB022433.1 |
- |
1, 2 |
162 |
|
AB209278.1 |
- |
1, 2 |
148 |
|
AF216389.1 |
- |
1, 2 |
127 |
|
AF293363.1 |
- |
1, 2 |
127 |
|
AK300037.1 |
- |
2, 3 |
386 |
|
AY358939.1 |
- |
1, 2 |
142 |
|
BC142617.1 |
- |
2, 3 |
329 |
40147342 |
DA818632.1 |
- |
2, 3 |
386 |
|
DB257858.1 |
- |
2, 3 |
436 |
|
FR839673.1 |
- |
1, 2 |
124 |
|
View Full Product Details