Gene Transcripts

Gene Symbol
PABPC5
Entrez Gene ID
140886
Gene Name
poly(A) binding protein cytoplasmic 5
Gene Aliases
PABP5
Chromosome Location
Chr. X: 91434598 - 91438584 on Build GRCh38
UniGene ID
Hs.246590
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080832.2 NP_543022.1 2 883
GenBank mRNA AK290105.1 - 1 448
AK297327.1 - 3 556
AL122118.1 - 1 398
BC063113.1 - 2 661 6452933

View Full Product Details