Gene Transcripts

Gene Symbol
CPNE4
Entrez Gene ID
131034
Gene Name
copine 4
Gene Aliases
COPN4, CPN4
Chromosome Location
Chr. 3: 131533560 - 132285702 on Build GRCh38
UniGene ID
Hs.199877
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289112.1 NP_001276041.1 3 452
NM_130808.2 NP_570720.1 3 1080
NM_153429.1 NP_702907.1 4 440
XM_011512406.2 XP_011510708.1 2 227
XM_011512407.2 XP_011510709.1 2 484
XM_011512408.2 XP_011510710.1 2 1612
XM_017005693.1 XP_016861182.1 6 1833
XM_017005694.1 XP_016861183.1 2 204
XM_017005695.1 XP_016861184.1 3 473
GenBank mRNA AF465770.1 - 6 780
AF465771.1 - 4 440
AK056175.1 - 3 552
AK128117.1 - 7 2330
AK295557.1 - 3 367
BC014396.1 - 3 638 3162811
BC075046.2 - 3 229 30915576
DC330179.1 - 3 452

View Full Product Details