Gene Transcripts

Gene Symbol
NUP35
Entrez Gene ID
129401
Gene Name
nucleoporin 35
Gene Aliases
MP-44, MP44, NP44, NUP53
Chromosome Location
Chr. 2: 183117490 - 183161684 on Build GRCh38
UniGene ID
Hs.180591
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001287584.1 NP_001274513.1 3 299
NM_001287585.1 NP_001274514.1 2 196
NM_138285.4 NP_612142.2 2 196
NR_109856.1 2 196
XM_006712254.2 XP_006712317.1 2 161
XM_011510576.2 XP_011508878.1 4 427
XM_011510577.2 XP_011508879.1 4 614
XM_017003308.1 XP_016858797.1 2 352
GenBank mRNA AF411516.1 - 2 196
AF514993.1 - 2 153
AK289601.1 - 2 139
AK290760.1 - 2 108
AK298199.1 - 2 106
AK302350.1 - 3 268
BC035597.1 - 2 120
BC047029.1 - 2 183 5541023
BC061698.1 - 2 150 4132440
BI818393.1 - 2 175
BI821231.1 - 2 133
CX787176.1 - 2 94
HY031440.1 - 4 443

View Full Product Details