Gene Transcripts

Gene Symbol
NUP35
Entrez Gene ID
129401
Gene Name
nucleoporin 35
Gene Aliases
MP-44, MP44, NP44, NUP53
Chromosome Location
Chr. 2: 183117490 - 183161684 on Build GRCh38
UniGene ID
Hs.180591
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001287584.1 NP_001274513.1 4 518
NM_001287585.1 NP_001274514.1 3 415
NM_138285.4 NP_612142.2 3 415
NR_109856.1 3 415
XM_006712254.2 XP_006712317.1 3 380
XM_011510576.2 XP_011508878.1 5 646
XM_011510577.2 XP_011508879.1 5 833
XM_017003308.1 XP_016858797.1 3 571
GenBank mRNA AF411516.1 - 3 415
AF514993.1 - 3 372
AK289601.1 - 3 358
AK290760.1 - 3 327
AK298199.1 - 3 325
AK302350.1 - 4 487
BC035597.1 - 3 339
BC047029.1 - 3 402 5541023
BC061698.1 - 3 369 4132440
BI818393.1 - 3 394
BI821231.1 - 3 351
CX787176.1 - 3 313

View Full Product Details