Gene Transcripts

Gene Symbol
MUC4
Entrez Gene ID
4585
Gene Name
mucin 4, cell surface associated
Gene Aliases
ASGP, HSA276359, MUC-4
Chromosome Location
Chr. 3: 195746765 - 195811973 on Build GRCh38
UniGene ID
Hs.369646
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001322468.1 NP_001309397.1 19621
NM_004532.5 NP_004523.3 6 895
NM_018406.6 NP_060876.5 13603
NM_138297.4 NP_612154.2 5 742
GenBank mRNA AJ010901.1 - 6 679
AJ242541.1 - 7 904
AJ242542.1 - 6 693
AJ242543.1 - 6 603
AJ242544.1 - 6 603
AJ242545.1 - 6 603
AJ242546.1 - 6 679
AJ242547.1 - 6 679
AJ242548.1 - 6 679
AJ242549.1 - 6 779
AJ242550.1 - 5 626
AJ276359.1 - 8 3789
AJ277412.1 - 7 3633
AJ277505.1 - 7 3619
AJ400633.1 - 8 3803
AJ400849.1 - 8 3713
AJ400850.1 - 8 3713
AJ400851.1 - 8 3803
AJ400852.1 - 7 3557
AJ400853.1 - 7 3557
AJ400854.1 - 7 3634
AJ400855.1 - 7 3633
AJ400856.1 - 8 3789
AJ400857.1 - 8 3789
AJ400858.1 - 8 3789
DB227644.1 - 4 489
EF091824.1 - 9 3664

View Full Product Details